3-Carboxy-2-methoxyphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Borono-2-methoxybenzoic acid, 3-Carboxy-2-methoxybenzeneboronic acid
H53323.03
H53323.03EA
174
EUR
H53323.03
3-Carboxy-2-methoxyphenylboronsäure ≥98%
3-Carboxy-2-methoxyphenylboronsäure
Purity: 98%
CAS No.: 913836-10-9
Molecular Formula: C8H9BO5
Formula Weight: 195.97
Storage Temperature: 2°C to 8°C
Melting Point: 152-155°
MDL No.: MFCD09027237
Formel:
C₈H₉BO₅ Molecular Weight: 195,97 g/mol Schmelzpunkt: 152…155 °C |
MDL:
MFCD09027237 CAS-Nummer: 913836-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...