4-Chlor-3-fluorphenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Chlor-3-fluorphenyl)metaborsäure, Dihydroxy(4-chlor-3-fluorphenyl)boran, 4-Chlor-3-fluorbenzolboronsäure

H53325.06
H53325.06EA 89.8 EUR
H53325.06
4-Chlor-3-fluorphenylboronsäure ≥97%
4-Chlor-3-fluorphenylboronsäure

Purity: 97% CAS No.: 137504-86-0 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 Melting Point: 198-204° MDL No.: MFCD01319010

Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
Schmelzpunkt: 198…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319010
CAS-Nummer: 137504-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR