3-(Cyanomethyl)benzeneboronic acid 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (3-(Cyanomethyl)phenyl)boronic acid

H53337.MD
H53337.MDEA 71 EUR
H53337.MD
3-(Cyanomethyl)benzeneboronic acid 96%
3-(Cyanomethyl)benzeneboronic acid

Purity: 96% CAS No.: 220616-39-7 Molecular Formula: C8H8BNO2 Formula Weight: 160.97 Melting Point: 85-88° MDL No.: MFCD03788026

Formel: C₈H₈BNO₂
Molecular Weight: 160,97 g/mol
Schmelzpunkt: 85…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788026
CAS-Nummer: 220616-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR