3,4-Dimethoxy-L-phenylalanine 97%

Lieferant: Thermo Scientific
Achtung

H53368.03
H53368.03EA 92 EUR
H53368.03
3,4-Dimethoxy-L-phenylalanine 97%
3,4-Dimethoxy-L-phenylalanine

Purity: 97% CAS No.: 32161-30-1 Molecular Formula: C11H15NO4 Formula Weight: 225.24 Melting Point: 254-257° Optical Rotation: -5° (c=4 in 1M hydrochloric acid) MDL No.: MFCD01321274

Formel: C₁₁H₁₅NO₄
Molecular Weight: 225,24 g/mol
Schmelzpunkt: 254…257 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01321274
CAS-Nummer: 32161-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR