N-(p-Toluenesulphonyl)glycine 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Tosylglycine, N-p-Tosylglycine, Tos-Gly-OH

H53371.06 H53371.14
H53371.06EA 49.4 EUR
H53371.06 H53371.14
N-(p-Toluenesulphonyl)glycine 97%
N-(p-Toluenesulphonyl)glycine

Purity: 97% CAS No.: 1080-44-0 Molecular Formula: C9H11NO4S Formula Weight: 229.25 Melting Point: 146-149° MDL No.: MFCD00045898

Formel: C₉H₁₁NO₄S
Molecular Weight: 229.25 g/mol
Schmelzpunkt: 146…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045898
CAS-Nummer: 1080-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR