N-(p-Toluenesulphonyl)glycine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Tosylglycine, N-p-Tosylglycine, Tos-Gly-OH
H53371.06
H53371.14
H53371.06EA
49.4
EUR
H53371.06
H53371.14
N-(p-Toluenesulphonyl)glycine 97%
N-(p-Toluenesulphonyl)glycine
Purity: 97%
CAS No.: 1080-44-0
Molecular Formula: C9H11NO4S
Formula Weight: 229.25
Melting Point: 146-149°
MDL No.: MFCD00045898
Formel:
C₉H₁₁NO₄S Molecular Weight: 229.25 g/mol Schmelzpunkt: 146…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045898 CAS-Nummer: 1080-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...