1-(1-Adamantyl)ethylamine hydrochloride 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Rimantadine Hydrochloride, alpha-Methyl-1-adamantanemethylamine Hydrochloride

H53377.06 H53377.14
H53377.06EA 87.4 EUR
H53377.06 H53377.14
1-(1-Adamantyl)ethylamine hydrochloride 98%
1-(1-Adamantyl)ethylamine hydrochloride

Purity: 99% CAS No.: 1501-84-4 Molecular Formula: C12H21N.HCl Formula Weight: 215.77 Melting Point: 375° MDL No.: MFCD00072023

Formel: C₁₂H₂₁N
Molecular Weight: 215.77 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00072023
CAS-Nummer: 1501-84-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR