6-Chlor-1-hexen ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H53396.06
H53396.14
H53396.06EA
67.7
EUR
H53396.06
H53396.14
6-Chlor-1-hexen ≥97%
6-Chlor-1-hexen
Purity: 97%
CAS No.: 928-89-2
Molecular Formula: C6H11Cl
Formula Weight: 118.61
Flash Point: 29°(84°F)
Refractive Index: 1.438
Boiling Point: 135-136°
UN No.: 1993
MDL No.: MFCD00039391
Formel:
C₆H₁₁Cl Siedepunkt: 135…136 °C Dichte: 0,896 Flammpunkt: 29 °C (84 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039391 CAS-Nummer: 928-89-2 EINECS: 213-186-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...