(2-Bromophenylethynyl)trimethylsilane 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98%
CAS No.: 38274-16-7
Molecular Formula: C11H13BrSi
Formula Weight: 253.21
Flash Point:
Formel:
C₁₁H₁₃BrSi Siedepunkt: 60…80 °C (2 mmHg) Dichte: 1,19 Flammpunkt: >113 °C (235 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01321391 CAS-Nummer: 38274-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...