(2-Bromophenylethynyl)trimethylsilane 98%

Lieferant: Thermo Fisher Scientific
Achtung

H53402.03 H53402.06 H53402.14
H53402.03EA 32.8 EUR
H53402.03 H53402.06 H53402.14
(2-Bromophenylethynyl)trimethylsilane 98%
(2-Bromophenylethynyl)trimethylsilane

Purity: 98% CAS No.: 38274-16-7 Molecular Formula: C11H13BrSi Formula Weight: 253.21 Flash Point: 113°(235°F) Physical Form: Liquid Density: 1.19 Refractive Index: 1.554 Appearance: Yellow to brown Boiling Point: 60-80°/2mm MDL No.: MFCD01321391

Formel: C₁₁H₁₃BrSi
Siedepunkt: 60…80 °C (2 mmHg)
Dichte: 1,19
Flammpunkt: >113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD01321391
CAS-Nummer: 38274-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR