(1S,2R,3R,4S)-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
cis-endo-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
H53403.14
H53403.14EA
141
EUR
H53403.14
(1S,2R,3R,4S)-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid 98%
(1S,2R,3R,4S)-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
Purity: 98%
CAS No.: 3853-88-1
Molecular Formula: C9H10O4
Formula Weight: 182.18
Melting Point: 175° dec.
MDL No.: MFCD00003735
Formel:
C₉H₁₀O₄ Molecular Weight: 182,18 g/mol Schmelzpunkt: 175 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026879 CAS-Nummer: 3853-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...