2,9-Dimethyl-5-decin 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H53414.03
H53414.03EA
71.5
EUR
H53414.03
2,9-Dimethyl-5-decin 96%
2,9-Dimethyl-5-decyne
Purity: 96%
CAS No.: 19550-56-2
Molecular Formula: C12H22
Formula Weight: 166.31
Boiling Point: 92°/19mm
MDL No.: MFCD00041611
Formel:
C₁₂H₂₂ Siedepunkt: 92 °C (19 mmHg) Dichte: 0,77 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041611 CAS-Nummer: 19550-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...