sec-Butyl methyl ether 99%

Lieferant: Thermo Fisher Scientific
Gefahr

H53427.AA H53427.AC H53427.AE
H53427.AAEA 44.7 EUR
H53427.AA H53427.AC H53427.AE
sec-Butyl methyl ether 99%
sec-Butyl methyl ether

Purity: 99% CAS No.: 6795-87-5 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: -30°(-22°F) Refractive Index: 1.372 Boiling Point: 57° UN No.: UN2350 MDL No.: MFCD00190205

Formel: C₅H₁₂O
Siedepunkt: 57 °C
Dichte: 0,742
Flammpunkt: –30 °C (–22 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00190205
CAS-Nummer: 6795-87-5
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR