N-(4-Pentinyl)phthalimid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Pent-4-yn-1-yl)isoindoline-1,3-dione

H53432.06 H53432.14
H53432.06EA 71.3 EUR
H53432.06 H53432.14
N-(4-Pentinyl)phthalimid ≥97%
N-(4-Pentinyl)phthalimid

Purity: 97% CAS No.: 6097-07-0 Molecular Formula: C13H11NO2 Formula Weight: 213.24 Melting Point: 87-91° MDL No.: MFCD06798110

Formel: C₁₃H₁₁NO₂
Schmelzpunkt: 87…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06798110
CAS-Nummer: 6097-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR