1-Chlor-2-octin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H53452.06
H53452.06EA 58 EUR
H53452.06
1-Chlor-2-octin ≥98%
1-Chlor-2-octin

Purity: 98% CAS No.: 51575-83-8 Molecular Formula: C8H13Cl Formula Weight: 144.64 Flash Point: 68°(154°F) Refractive Index: 1.46 Boiling Point: 81°/15mm MDL No.: MFCD00274223

Formel: C₈H₁₃Cl
Molecular Weight: 144.64 g/mol
Siedepunkt: 81 °C (15 mmHg)
Dichte: 0.931
Flammpunkt: 68 °C (154 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00274223
CAS-Nummer: 51575-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR