6-Chlor-2-hexanon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H53462.06
H53462.14
H53462.06EA
68.8
EUR
H53462.06
H53462.14
6-Chlor-2-hexanon ≥98%
6-Chlor-2-hexanon
Purity: 98%
CAS No.: 10226-30-9
Molecular Formula: C6H11ClO
Formula Weight: 134.61
Flash Point: 91°(196°F)
Refractive Index: 1.4435
Boiling Point: 86°/16mm
MDL No.: MFCD00191625
Formel:
C₆H₁₁ClO Siedepunkt: 86 °C (16 mmHg) Dichte: 1,02 Flammpunkt: 91 °C (196 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191625 CAS-Nummer: 10226-30-9 EINECS: 233-546-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...