Propargyl ether 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H53473.06
H53473.14
H53473.06EA
109
EUR
H53473.06
H53473.14
Propargyl ether 98%
Propargyl ether
Purity: 98%
CAS No.: 6921-27-3
Molecular Formula: C6H6O
Formula Weight: 94.11
Flash Point: 24°(75°F)
Refractive Index: 1.4430
Boiling Point: 55-57°/65mm
UN No.: UN3271
MDL No.: MFCD00048108
Formel:
C₆H₆O Siedepunkt: 55…57 °C (65 mmHg) Dichte: 0,914 Flammpunkt: 24 °C (75 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048108 CAS-Nummer: 6921-27-3 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...