2-(Trifluoromethyl)phenylacetylene 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H53478.03
H53478.06
H53478.03EA
53.9
EUR
H53478.03
H53478.06
2-(Trifluoromethyl)phenylacetylene 97%
2-(Trifluoromethyl)phenylacetylene
Purity: 97%
CAS No.: 704-41-6
Molecular Formula: C9H5F3
Formula Weight: 170.13
Flash Point: 25°(77°F)
Refractive Index: 1.4760
Boiling Point: 45°/8mm
UN No.: UN1993
MDL No.: MFCD00467313
Formel:
C₉H₅F₃ Siedepunkt: 45 °C (8 mmHg) Dichte: 1,222 Flammpunkt: 25 °C (77 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00467313 CAS-Nummer: 704-41-6 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...