cis-3-Hexen ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H53485.06
H53485.14
H53485.06EA
89.2
EUR
H53485.06
H53485.14
cis-3-Hexen ≥97%
cis-3-Hexene
Purity: 97%
CAS No.: 7642-09-3
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -25°(-13°F)
Refractive Index: 1.395
Boiling Point: 66-68°
Beilstein Reference: 1718858
UN No.: 3295
MDL No.: MFCD00085300
Formel:
C₆H₁₂ Siedepunkt: 66…68 °C Dichte: 0,681 Flammpunkt: –25 °C (–13 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085300 CAS-Nummer: 7642-09-3 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...