N-(5-Hexinyl)phthalimid ≥97%

Lieferant: Thermo Fisher Scientific

H53489.06 H53489.14
H53489.06EA 82.2 EUR
H53489.06 H53489.14
N-(5-Hexinyl)phthalimid ≥97%
N-(5-Hexinyl)phthalimid

Purity: 97% CAS No.: 6097-08-1 Molecular Formula: C14H13NO2 Formula Weight: 227.26 Melting Point: 68-72° MDL No.: MFCD00671372

Formel: C₁₄H₁₃NO₂
Schmelzpunkt: 68…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00671372
CAS-Nummer: 6097-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR