3-Ethyl-3-oxetanmethanol ≥97%

Lieferant: Thermo Fisher Scientific

H53497.06 H53497.14
H53497.06EA 35.1 EUR
H53497.06 H53497.14
3-Ethyl-3-oxetanmethanol ≥97%
3-Ethyl-3-oxetanmethanol

Purity: 97% CAS No.: 3047-32-3 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 108°(226°F) Refractive Index: 1.453 Boiling Point: 96°/4mm MDL No.: MFCD00190143

Formel: C₆H₁₂O₂
Siedepunkt: 96 °C (4 mmHg)
Dichte: 1,019
Flammpunkt: 108 °C (226 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00190143
CAS-Nummer: 3047-32-3
EINECS: 221-254-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR