2-[Di(methylthio)methylidene]malononitrile 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(Bis(methylthio)methylene)malononitrile

H53503.14
H53503.14EA 251 EUR
H53503.14
2-[Di(methylthio)methylidene]malononitrile 97%
2-[Di(methylthio)methylidene]malononitrile

Purity: 97% CAS No.: 5147-80-8 Molecular Formula: C6H6N2S2 Formula Weight: 170.25 Melting Point: 79-82° UN No.: UN3439 MDL No.: MFCD00052730

Formel: C₆H₆N₂S₂
Molecular Weight: 170,26 g/mol
Schmelzpunkt: 79…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052730
CAS-Nummer: 5147-80-8
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR