cis-2-Hexen 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H53506.06
H53506.14
H53506.06EA
68.8
EUR
H53506.06
H53506.14
cis-2-Hexen 96%
cis-2-Hexen
Purity: 96%
CAS No.: 7688-21-3
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -25°(-13°F)
Refractive Index: 1.396
Boiling Point: 68-70°
Beilstein Reference: 1719019
UN No.: 3295
MDL No.: MFCD00066520
Formel:
C₆H₁₂ Siedepunkt: 68…70 °C Dichte: 0,669 Flammpunkt: –25 °C (–13 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066520 CAS-Nummer: 7688-21-3 EINECS: 231-697-1 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...