3-Chlor-1-phenyl-1-propin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (3-Chlor-1-propinyl)benzol

H53510.14 H53510.03 H53510.06
H53510.14EA 397 EUR
H53510.14 H53510.03 H53510.06
3-Chlor-1-phenyl-1-propin ≥97%
3-Chlor-1-phenyl-1-propin

Purity: 97% CAS No.: 3355-31-5 Molecular Formula: C9H7Cl Formula Weight: 150.61 Flash Point: 104°(219°F) Refractive Index: 1.585 MDL No.: MFCD06411085

Formel: C₉H₇Cl
Molecular Weight: 150.61 g/mol
Dichte: 1.095
Flammpunkt: 104 °C (219 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD06411085
CAS-Nummer: 3355-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR