O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate

H54035.06
H54035.06EA 92.5 EUR
H54035.06
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate ≥98%
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate

Purity: 98+% CAS No.: 265651-18-1 Molecular Formula: C9H16F6N3O3P Formula Weight: 359.21 Storage Temperature: -30°C to -10°C Melting Point: ca 220° dec. MDL No.: MFCD01863753

Formel: C₉H₁₆F₆N₃O₃P
Molecular Weight: 359,21 g/mol
Schmelzpunkt: ∼ 220 °C (Zersetzung)
MDL: MFCD01863753
CAS-Nummer: 265651-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR