5-Bromo-2-[3-(chloromethyl)phenyl]pyrimidine 97%

Lieferant: Thermo Scientific
Achtung

H54135.06
H54135.06EA 497 EUR
H54135.06
5-Bromo-2-[3-(chloromethyl)phenyl]pyrimidine 97%
5-Bromo-2-[3-(chloromethyl)phenyl]pyrimidine

Purity: 97% CAS No.: 1100598-50-2 Molecular Formula: C11H8BrClN2 Formula Weight: 283.55 MDL No.: MFCD20527211

Formel: C₁₁H₈BrClN₂
Molecular Weight: 283,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20527211
CAS-Nummer: 1100598-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR