O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,1,3,3-Tetramethyl-2-(4-oxobenzo[d][1,2,3]triazin-3(4H)-yl)isouronium tetrafluoroborate, N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium Tetrafluoroborate, TDBTU

H54144.06
H54144.06EA 106 EUR
H54144.06
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98%
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate

Purity: 98+% CAS No.: 125700-69-8 Molecular Formula: C12H16BF4N5O2 Formula Weight: 349.09 Storage Temperature: 2°C to 8°C MDL No.: MFCD00077730

Formel: C₁₂H₁₆BF₄N₅O₂
Molecular Weight: 349,10 g/mol
MDL: MFCD00077730
CAS-Nummer: 125700-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR