1-Methyl-1H-pyrazol-4-carbaldehyd 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Methyl-1H-pyrazole-4-carbaldehyde, 1-Methyl-1H-pyrazole-4-carboxaldehyde

H54146.03
H54146.03EA 80.9 EUR
H54146.03
1-Methyl-1H-pyrazol-4-carbaldehyd 96%
1-Methyl-1H-pyrazol-4-carbaldehyd

Purity: 96% CAS No.: 25016-11-9 Molecular Formula: C5H6N2O Formula Weight: 110.12 Flash Point: 230°(446°F) Melting Point: 29-34° MDL No.: MFCD00460465

Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Schmelzpunkt: 29…34 °C
Flammpunkt: >230 °C (446 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00460465
CAS-Nummer: 25016-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR