2,6-Difluorobenzylmagnesium bromide 0.25 M in 2-MeTHF (2-Methyltetrahydrofuran)

Lieferant: Thermo Fisher Scientific
Gefahr

H54189.AE
H54189.AEEA 238 EUR
H54189.AE
2,6-Difluorobenzylmagnesium bromide 0.25 M in 2-MeTHF (2-Methyltetrahydrofuran)
2,6-Difluorobenzylmagnesium bromide

CAS No.: 107549-26-8 Molecular Formula: C7H5BrF2Mg Formula Weight: 231.32 UN No.: UN2924 MDL No.: MFCD11113508

Formel: C₇H₅BrF₂Mg
Lagertemperatur: Raumtemperatur
MDL: MFCD11113508
CAS-Nummer: 107549-26-8
UN: 2924
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR