2-Hydroxypyridin-N-oxid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H54207.06
H54207.14
H54207.22
H54207.06EA
28.3
EUR
H54207.06
H54207.14
H54207.22
2-Hydroxypyridin-N-oxid ≥98%
2-Hydroxypyridin-N-oxid
Purity: 98+%
CAS No.: 13161-30-3
Molecular Formula: C5H5NO2
Formula Weight: 111.10
Flash Point: 100°(212°F)
Melting Point: 147-152°
Beilstein Reference: 1422545
MDL No.: MFCD00006195
Formel:
C₅H₅NO₂ Schmelzpunkt: 147…152 °C Dichte: 1,111 Flammpunkt: 100 °C (212 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006195 CAS-Nummer: 13161-30-3 EINECS: 236-100-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...