4-(Hydroxymethyl)benzonitril ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Cyanobenzyl alcohol

H54283.03 H54283.06
H54283.03EA 67.3 EUR
H54283.03 H54283.06
4-(Hydroxymethyl)benzonitril ≥97%
4-(Hydroxymethyl)benzonitril

Purity: 97% CAS No.: 874-89-5 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 32-36° UN No.: 3439 MDL No.: MFCD00870633

Formel: C₈H₇NO
Schmelzpunkt: 32…36 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00870633
CAS-Nummer: 874-89-5
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR