4-(Hydroxymethyl)benzonitril ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Cyanobenzyl alcohol
H54283.03
H54283.06
H54283.03EA
67.3
EUR
H54283.03
H54283.06
4-(Hydroxymethyl)benzonitril ≥97%
4-(Hydroxymethyl)benzonitril
Purity: 97%
CAS No.: 874-89-5
Molecular Formula: C8H7NO
Formula Weight: 133.15
Melting Point: 32-36°
UN No.: 3439
MDL No.: MFCD00870633
Formel:
C₈H₇NO Schmelzpunkt: 32…36 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00870633 CAS-Nummer: 874-89-5 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...