4-tert-Butylphenylmagnesium bromide 0.50 M in 2-MeTHF (2-Methyltetrahydrofuran)
Lieferant: Thermo Scientific
|
Gefahr
|
H54284.AE
H54284.AEEA
191
EUR
H54284.AE
4-tert-Butylphenylmagnesium bromide 0.50 M in 2-MeTHF (2-Methyltetrahydrofuran)
4-tert-Butylphenylmagnesium bromide
CAS No.: 63488-10-8
Molecular Formula: C10H13BrMg
Formula Weight: 237.42
UN No.: UN2924
MDL No.: MFCD00013244
Formel:
C₁₀H₁₃BrMg Molecular Weight: 237,42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013244 CAS-Nummer: 63488-10-8 UN: 2924 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...