3-Amino-5-brom-2-methoxypyridin 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Bromo-2-methoxypyridin-3-amine

H54299.06
H54299.06EA 524 EUR
H54299.06
3-Amino-5-brom-2-methoxypyridin 96%
3-Amino-5-bromo-2-methoxypyridine

Purity: 96% CAS No.: 884495-39-0 Molecular Formula: C6H7BrN2O Formula Weight: 203.04 Melting Point: 53-55° MDL No.: MFCD07368887

Formel: C₆H₇BrN₂O
Molecular Weight: 203,04 g/mol
Schmelzpunkt: 53…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368887
CAS-Nummer: 884495-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR