2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H54534.MD
H54534.03
H54534.MDEA
92.6
EUR
H54534.MD
H54534.03
2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester 96%
2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester
Purity: 96%
CAS No.: 1111637-92-3
Molecular Formula: C13H21BN2O2
Formula Weight: 248.13
Storage Temperature: 2°C to 8°C
Melting Point: 105-108°
MDL No.: MFCD12923387
Formel:
C₁₃H₂₁BN₂O₂ Schmelzpunkt: 105…108 °C |
MDL:
MFCD12923387 CAS-Nummer: 1111637-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...