Bis(2-methyl-2-propanyl)-[(Z)-1H-pyrazol-1-ylmethylyliden]biscarbamat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Bis-Boc-pyrazolocarboxamidine, 1-[N,N'-Bis(tert-butoxycarbonyl)carbamimidoyl]-1H-pyrazole, N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, 1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected, N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboximidamide

H54565.03 H54565.06 H54565.14
H54565.03EA 49 EUR
H54565.03 H54565.06 H54565.14
Bis(2-methyl-2-propanyl)-[(Z)-1H-pyrazol-1-ylmethylyliden]biscarbamat ≥98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate

Purity: 98+% CAS No.: 152120-54-2 Molecular Formula: C14H22N4O4 Formula Weight: 310.35 Melting Point: 86-90° MDL No.: MFCD01075122

Formel: C₁₄H₂₂N₄O₄
Schmelzpunkt: 86…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075122
CAS-Nummer: 152120-54-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR