1,3-Cycloheptandion 97%
Lieferant: Thermo Scientific
Synonyme:
1,3-Cycloheptanedione
H54588.03
H54588.03EA
169
EUR
H54588.03
1,3-Cycloheptandion 97%
Cycloheptane-1,3-dione
Purity: 97%
CAS No.: 1194-18-9
Molecular Formula: C7H10O2
Formula Weight: 126.15
Flash Point:
Formel:
C₇H₁₀O₂ Molecular Weight: 126,155 g/mol Siedepunkt: 254 °C Dichte: 1,1 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863735 CAS-Nummer: 1194-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...