1H-Indol-6-ylmethylamine 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(1H-Indol-6-yl)methanamine, 6-(Aminomethyl)indole
H54640.MD
H54640.03
H54640.MDEA
53.7
EUR
H54640.MD
H54640.03
1H-Indol-6-ylmethylamine 97%
1H-Indol-6-ylmethylamine
Purity: 97%
CAS No.: 3468-17-5
Molecular Formula: C9H10N2
Formula Weight: 146.19
MDL No.: MFCD06213839
Formel:
C₉H₁₀N₂ Molecular Weight: 146.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06213839 CAS-Nummer: 3468-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...