1H-Indol-6-ylmethylamine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (1H-Indol-6-yl)methanamine, 6-(Aminomethyl)indole

H54640.MD H54640.03
H54640.MDEA 53.7 EUR
H54640.MD H54640.03
1H-Indol-6-ylmethylamine 97%
1H-Indol-6-ylmethylamine

Purity: 97% CAS No.: 3468-17-5 Molecular Formula: C9H10N2 Formula Weight: 146.19 MDL No.: MFCD06213839

Formel: C₉H₁₀N₂
Molecular Weight: 146.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213839
CAS-Nummer: 3468-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR