1-Chloroacetyl-2-(trifluoroacetyl)hydrazine 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-(Chloroacetyl)-2-(trifluoroacetyl)hydrazine
H54647.MD
H54647.03
H54647.06
H54647.MDEA
43.3
EUR
H54647.MD
H54647.03
H54647.06
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine 97%
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine
Purity: 97%
CAS No.: 762240-99-3
Molecular Formula: C4H4ClF3N2O2
Formula Weight: 204.53
MDL No.: MFCD07779449
Formel:
C₄H₄ClF₃N₂O₂ Molecular Weight: 204.53 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779449 CAS-Nummer: 762240-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...