1-Chloroacetyl-2-(trifluoroacetyl)hydrazine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(Chloroacetyl)-2-(trifluoroacetyl)hydrazine

H54647.MD H54647.03 H54647.06
H54647.MDEA 43.3 EUR
H54647.MD H54647.03 H54647.06
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine 97%
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine

Purity: 97% CAS No.: 762240-99-3 Molecular Formula: C4H4ClF3N2O2 Formula Weight: 204.53 MDL No.: MFCD07779449

Formel: C₄H₄ClF₃N₂O₂
Molecular Weight: 204.53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779449
CAS-Nummer: 762240-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR