5-Aminopyridine-2-carboxamidoxime 97%
Lieferant: Thermo Scientific
|
Achtung
|
H54666.06
H54666.06EA
486
EUR
H54666.06
5-Aminopyridine-2-carboxamidoxime 97%
5-Aminopyridine-2-carboxamidoxime
Purity: 97%
Molecular Formula: C6H8N4O
Formula Weight: 152.16
MDL No.: MFCD20265317
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...