Butamben (Butyl-4-aminobenzoat) 99%

Lieferant: Thermo Fisher Scientific
Achtung

H54690.22 H54690.36 H54690.0B
H54690.22EA 67.2 EUR
H54690.22 H54690.36 H54690.0B
Butamben (Butyl-4-aminobenzoat) 99%
Butamben (Butyl-4-aminobenzoat)

Purity: 99% CAS No.: 94-25-7 Molecular Formula: C11H15NO2 Formula Weight: 193.25 Melting Point: 56-59° Boiling Point: 174°/8mm Merck Reference: 14,1512 MDL No.: MFCD00017112

Formel: C₁₁H₁₅NO₂
Siedepunkt: 174 °C (8 mmHg)
Schmelzpunkt: 56…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017112
CAS-Nummer: 94-25-7
EINECS: 202-317-1
Merck Index: 14,1512

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR