2-Amino-5-brom-3-methoxypyridin 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromo-3-methoxypyridin-2-amine

H54723.03
H54723.03EA 260 EUR
H54723.03
2-Amino-5-brom-3-methoxypyridin 96%
2-Amino-5-bromo-3-methoxypyridine

Purity: 96% CAS No.: 42409-58-5 Molecular Formula: C6H7BrN2O Formula Weight: 203.04 Melting Point: 78-82° MDL No.: MFCD09909849

Formel: C₆H₇BrN₂O
Molecular Weight: 203,04 g/mol
Schmelzpunkt: 78…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09909849
CAS-Nummer: 42409-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR