4-Chloro-2,6-dimethylpyrimidine 97%

Lieferant: Thermo Fisher Scientific
Achtung

H54735.06
H54735.06EA 784 EUR
H54735.06
4-Chloro-2,6-dimethylpyrimidine 97%
4-Chloro-2,6-dimethylpyrimidine

Purity: 97% CAS No.: 4472-45-1 Molecular Formula: C6H7ClN2 Formula Weight: 142.03 MDL No.: MFCD00234197

Formel: C₆H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00234197
CAS-Nummer: 4472-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR