6-Brom-4-methyl-3-nitrochinolin 96%
Lieferant: Thermo Scientific
|
Achtung
|
H54769.03
H54769.03EA
504
EUR
H54769.03
6-Brom-4-methyl-3-nitrochinolin 96%
6-Bromo-4-methyl-3-nitroquinoline
Purity: 96%
CAS No.: 1185310-49-9
Molecular Formula: C10H7BrN2O2
Formula Weight: 267.08
Storage Temperature: 2°C to 8°C
Melting Point: 152-155°
Beilstein Reference: 19927450
MDL No.: MFCD12761249
Formel:
C₁₀H₇BrN₂O₂ Molecular Weight: 267,08 g/mol Schmelzpunkt: 152…155 °C |
MDL:
MFCD12761249 CAS-Nummer: 1185310-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...