2-Methoxy-6-chinolinboronsäurepinakolester 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

H54814.03
H54814.03EA 240 EUR
H54814.03
2-Methoxy-6-chinolinboronsäurepinakolester 96%
2-Methoxy-6-chinolinboronsäurepinakolester

Purity: 96% Molecular Formula: C16H20BNO3 Formula Weight: 285.15 MDL No.: MFCD18383956

Formel: C₁₆H₂₀BNO₃
Molecular Weight: 285,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18383956
CAS-Nummer: 1201644-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR