tert-Pentylmagnesium chloride 1 M in 2-MeTHF (2-Methyltetrahydrofuran)

Lieferant: Thermo Fisher Scientific
Gefahr

H54824.AE
H54824.AEEA 436 EUR
H54824.AE
tert-Pentylmagnesium chloride 1 M in 2-MeTHF (2-Methyltetrahydrofuran)
tert-Pentylmagnesium chloride
Formel: C₅H₁₁ClMg
Dichte: 0,831
Lagertemperatur: Raumtemperatur
MDL: MFCD00010356
CAS-Nummer: 28276-08-6
UN: 3399
ADR: 4.3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR