Methyl-2-amino-3-brom-5-methoxybenzoat 96%

Lieferant: Thermo Scientific
Achtung

H54844.03
H54844.03EA 226 EUR
H54844.03
Methyl-2-amino-3-brom-5-methoxybenzoat 96%
Methyl-2-amino-3-brom-5-methoxybenzoat

Purity: 96% Molecular Formula: C9H10BrNO3 Formula Weight: 260.09 MDL No.: MFCD20527215

Formel: C₉H₁₀BrNO₃
Molecular Weight: 260,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20527215
CAS-Nummer: 1378874-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR