4-Fluoro-2-methylphenylmagnesium bromide 0.50 M in 2-MeTHF (2-Methyltetrahydrofuran)

Lieferant: Thermo Fisher Scientific
Gefahr

H54865.AE
H54865.AEEA 299 EUR
H54865.AE
4-Fluoro-2-methylphenylmagnesium bromide 0.50 M in 2-MeTHF (2-Methyltetrahydrofuran)

CAS No.: 30897-90-6 Molecular Formula: C7H6BrFMg Formula Weight: 213.33 UN No.: UN2924 MDL No.: MFCD01311469

Formel: C₇H₆BrFMg
Lagertemperatur: Raumtemperatur
MDL: MFCD01311469
CAS-Nummer: 30897-90-6
UN: 2924
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR